Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msi1
Entrez Gene ID
17690
Gene Name
musashi RNA-binding protein 1
Gene Aliases
Msi1h, Musahi1, m-Msi-1
Chromosome Location
Chr.5: 115359083 - 115455698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008629.1 NP_032655.1 6 - 7 462 70
GenBank mRNA AK084019.1 - 6 - 7 445 70
D49654.1 - 6 - 7 462 70

View Full Product Details