The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175309.4 | NP_780518.1 | 2 - 3 | 543 | 60 |
GenBank mRNA | AK042067.1 | - | 2 - 3 | 339 | 60 |
AK052525.1 | - | 2 - 3 | 545 | 60 | |
AK157647.1 | - | 2 - 3 | 350 | 60 | |
AY233463.1 | - | 2 - 3 | 337 | 60 | |
BC061224.1 | - | 2 - 3 | 340 | 60 | |
BC120594.1 | - | 2 - 3 | 293 | 60 | |
BC137742.1 | - | 2 - 3 | 293 | 60 |