Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Saal1
Entrez Gene ID
78935
Gene Name
serum amyloid A-like 1
Gene Aliases
5031425D22Rik, SPACIA1
Chromosome Location
Chr.7: 46686108 - 46710691 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030233.1 NP_084509.1 2 - 3 275 80
GenBank mRNA AB489137.1 - 2 - 3 256 80
AK019878.1 - 2 - 3 276 80
AK030833.1 - 2 - 3 281 80
AK031128.1 - 1 - 2 61 80
AK033202.1 - 2 - 3 286 80
AK132193.1 - 2 - 3 276 80
AK134418.1 - 2 - 3 283 80
BC068127.1 - 2 - 3 272 80
BC087944.1 - 2 - 3 287 80

View Full Product Details