Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scaf1
Entrez Gene ID
233208
Gene Name
SR-related CTD-associated factor 1
Gene Aliases
AI480556, SR-A1
Chromosome Location
Chr.7: 45002948 - 45016357 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK141920.1 - 2 - 3 186 84
BC043138.1 - 1 - 2 202 84

View Full Product Details