Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspb8
Entrez Gene ID
80888
Gene Name
heat shock protein 8
Gene Aliases
AU018630, AW413033, Cryac, D5Ucla4, E2IG1, H11, H11K, HSP20-like, HSP22
Chromosome Location
Chr.5: 116408495 - 116422864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF250139.1 - 2 - 3 451 73
AF273453.1 - 2 - 3 797 73
AK005052.1 - 2 - 3 819 73
BC003971.1 - 2 - 3 807 73
BC011219.1 - 2 - 3 800 73

View Full Product Details