Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt3
Entrez Gene ID
20981
Gene Name
synaptotagmin III
Gene Aliases
AI385753, sytIII
Chromosome Location
Chr.7: 44384099 - 44400030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114116.1 NP_001107588.1 4 - 5 888 99
NM_016663.3 NP_057872.3 4 - 5 915 99
GenBank mRNA AB000893.1 - 2 - 3 678 99
BC051969.1 - 3 - 4 796 99
D45858.1 - 3 - 4 849 99

View Full Product Details