Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt3
Entrez Gene ID
20981
Gene Name
synaptotagmin III
Gene Aliases
AI385753, sytIII
Chromosome Location
Chr.7: 44384099 - 44400030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114116.1 NP_001107588.1 9 - 10 1911 70
NM_016663.3 NP_057872.3 9 - 10 1938 70
GenBank mRNA AB000893.1 - 7 - 8 1701 70
BC051969.1 - 8 - 9 1819 70
D45858.1 - 8 - 9 1872 70

View Full Product Details