The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243052.1 | NP_001229981.1 | 3 - 4 | 549 | 90 |
NM_001243053.1 | NP_001229982.1 | 4 - 5 | 434 | 90 | |
NM_009737.3 | NP_033867.1 | 3 - 4 | 522 | 90 | |
GenBank mRNA | AF031467.1 | - | 3 - 4 | 314 | 90 |
AK037482.1 | - | 3 - 4 | 327 | 90 | |
AK137049.1 | - | 4 - 5 | 485 | 90 | |
AK145300.1 | - | 3 - 4 | 367 | 90 | |
AK160159.1 | - | 3 - 4 | 550 | 90 | |
BB630667.1 | - | 3 - 4 | 325 | 90 | |
BC017688.1 | - | 3 - 4 | 299 | 90 | |
BC048072.1 | - | 3 - 4 | 313 | 90 | |
U68526.1 | - | 3 - 4 | 308 | 90 |