Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E2f8
Entrez Gene ID
108961
Gene Name
E2F transcription factor 8
Gene Aliases
4432406C08Rik, AA410048
Chromosome Location
Chr.7: 48866429 - 48882142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013368.5 NP_001013386.2 3 - 4 788 76
GenBank mRNA AK049525.1 - 3 - 4 755 76
AK084513.1 - 3 - 4 789 76
AK154018.1 - 3 - 4 746 76
AK157235.1 - 3 - 4 748 76
AK160240.1 - 3 - 4 749 76
AY957576.1 - 3 - 4 598 76
BC086675.1 - 2 - 3 310 76

View Full Product Details