Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2f8
Entrez Gene ID
108961
Gene Name
E2F transcription factor 8
Gene Aliases
4432406C08Rik, AA410048
Chromosome Location
Chr.7: 48866429 - 48882142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013368.5 NP_001013386.2 4 - 5 940 76
GenBank mRNA AK049525.1 - 4 - 5 907 76
AK154018.1 - 4 - 5 898 76
AK157235.1 - 4 - 5 900 76
AK160240.1 - 4 - 5 901 76
AY957576.1 - 4 - 5 750 76

View Full Product Details