Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prune2
Entrez Gene ID
353211
Gene Name
prune homolog 2 (Drosophila)
Gene Aliases
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367
Chromosome Location
Chr.19: 16955670 - 17223958 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181348.4 NP_851993.3 2 - 3 282 89
GenBank mRNA AK020727.1 - 2 - 3 283 89
AK039241.1 - 2 - 3 283 89
AK165913.1 - 2 - 3 283 89

View Full Product Details