Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd10
Entrez Gene ID
330171
Gene Name
potassium channel tetramerisation domain containing 10
Gene Aliases
AW536343, C87062, mBACURD3
Chromosome Location
Chr.5: 114363572 - 114380532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159941.1 NP_001153413.1 2 - 3 294 61
NM_026145.4 NP_080421.2 2 - 3 294 61
GenBank mRNA AK028657.1 - 2 - 3 261 61
AK041516.1 - 2 - 3 261 61
AK145295.1 - 2 - 3 273 61
AK147714.1 - 2 - 3 239 61
AK161199.1 - 2 - 3 251 61
AK163351.1 - 2 - 3 248 61
AK166505.1 - 2 - 3 273 61
BC006935.1 - 2 - 3 275 61
BQ571808.1 - 2 - 3 245 61

View Full Product Details