The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029956.3 | NP_084232.1 | 8 - 9 | 669 | 64 |
GenBank mRNA | AK020286.1 | - | 8 - 9 | 669 | 64 |
AK088785.1 | - | 8 - 9 | 661 | 64 | |
AK148421.1 | - | 8 - 9 | 704 | 64 | |
BC022159.1 | - | 8 - 9 | 623 | 64 | |
BC057558.1 | - | 8 - 9 | 669 | 64 |