Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrif1
Entrez Gene ID
321000
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
2010012G17Rik, 4933421E11Rik, AI450568
Chromosome Location
Chr.3: 106684934 - 106736577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039488.1 NP_001034577.1 2 - 3 1906 149

View Full Product Details