The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293689.1 | NP_001280618.1 | 1 - 2 | 412 | 88 |
NM_172271.2 | NP_758475.1 | 1 - 2 | 412 | 88 | |
GenBank mRNA | AK083807.1 | - | 1 - 2 | 313 | 88 |
AK163113.1 | - | 1 - 2 | 413 | 88 | |
AY155578.1 | - | 1 - 2 | 413 | 88 |