The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011838.4 | NP_035968.1 | 3 - 4 | 380 | 62 |
GenBank mRNA | AF141377.1 | - | 3 - 4 | 354 | 62 |
AK013827.1 | - | 3 - 4 | 377 | 62 | |
AK161907.1 | - | 3 - 4 | 410 | 62 | |
AK162920.1 | - | 3 - 4 | 403 | 62 | |
AK165295.1 | - | 3 - 4 | 384 | 62 | |
AK165314.1 | - | 3 - 4 | 384 | 62 | |
BC037541.1 | - | 3 - 4 | 427 | 62 |