Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smurf1
Entrez Gene ID
75788
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases
4930431E10Rik, mKIAA1625
Chromosome Location
Chr.5: 144876495 - 144965830 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038627.1 NP_001033716.1 2 - 3 373 66
NM_029438.3 NP_083714.3 2 - 3 373 66
GenBank mRNA AK154491.1 - 2 - 3 374 66
BC029097.1 - 2 - 3 401 66

View Full Product Details