The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134152.3 | NP_598913.1 | 7 - 8 | 794 | 74 |
GenBank mRNA | AB053936.2 | - | 7 - 8 | 846 | 74 |
AB071194.1 | - | 7 - 8 | 819 | 74 | |
AK077085.1 | - | 7 - 8 | 830 | 74 | |
BC026563.1 | - | 7 - 8 | 760 | 74 |