The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267622.1 | NP_001254551.1 | 23 - 23 | 9644 | 138 |
GenBank mRNA | AK132014.1 | - | 1 - 1 | 251 | 138 |
AK172165.1 | - | 1 - 1 | 82 | 138 | |
BC002262.1 | - | 1 - 1 | 528 | 138 |