Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucb1
Entrez Gene ID
18220
Gene Name
nucleobindin 1
Gene Aliases
B230337F23Rik, C77483, Calnuc, MTEST82, Nucb
Chromosome Location
Chr.7: 45492674 - 45510408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163662.1 NP_001157134.1 4 - 5 468 68
NM_008749.2 NP_032775.1 4 - 5 468 68
GenBank mRNA AK002630.1 - 4 - 5 449 68
AK004886.1 - 4 - 5 439 68
AK007976.1 - 4 - 5 442 68
AK045087.1 - 4 - 5 468 68
AK140710.1 - 4 - 5 446 68
AK160981.1 - 4 - 5 455 68
AK164088.1 - 4 - 5 450 68
AK170717.1 - 4 - 5 463 68
BC072554.1 - 4 - 5 446 68
M96823.1 - 3 - 4 390 68

View Full Product Details