Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ruvbl2
Entrez Gene ID
20174
Gene Name
RuvB-like protein 2
Gene Aliases
mp47, p47
Chromosome Location
Chr.7: 45421896 - 45435170 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011304.3 NP_035434.1 11 - 12 1209 92
GenBank mRNA AB013912.1 - 11 - 12 1040 92
AK135404.1 - 11 - 12 1023 92
AK144156.1 - 11 - 12 1031 92
AK159112.1 - 11 - 12 1211 92
BC072585.1 - - 1708 92
BC115809.1 - 11 - 12 1027 92
BC116693.1 - 11 - 12 1026 92

View Full Product Details