The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007527.3 | NP_031553.1 | 1 - 2 | 159 | 81 |
GenBank mRNA | AK013298.1 | - | 1 - 2 | 69 | 81 |
AK149994.1 | - | 1 - 2 | 52 | 81 | |
AK159233.1 | - | 1 - 2 | 124 | 81 | |
BC018228.1 | - | 1 - 2 | 104 | 81 | |
BC053380.1 | - | 1 - 2 | 76 | 81 | |
BY308683.1 | - | 1 - 2 | 139 | 81 |