Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fut1
Entrez Gene ID
14343
Gene Name
fucosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.7: 45617266 - 45621059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271981.1 NP_001258910.1 2 - 2 645 91
NM_008051.6 NP_032077.2 2 - 2 783 91
GenBank mRNA AF113533.1 - 1 - 1 639 91
AK137638.1 - 2 - 2 785 91
BC109145.1 - 2 - 2 621 91
BC109146.1 - 2 - 2 621 91

View Full Product Details