Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tulp2
Entrez Gene ID
56734
Gene Name
tubby-like protein 2
Gene Aliases
Pdet
Chromosome Location
Chr.7: 45482026 - 45523712 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF105712.1 - 5 - 6 398 116
AK054304.1 - 2 - 3 356 116
AK077200.1 - 5 - 6 407 116
AK161165.1 - 2 - 3 355 116
BB617192.1 - 5 - 6 407 116

View Full Product Details