The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253814.1 | NP_001240743.1 | 10 - 11 | 2149 | 61 |
NM_027922.2 | NP_082198.1 | 10 - 11 | 2146 | 61 | |
GenBank mRNA | AK003234.1 | - | 7 - 8 | 948 | 61 |
AK129192.1 | - | 9 - 10 | 1475 | 61 | |
AK132187.1 | - | 10 - 11 | 2151 | 61 | |
AK142497.1 | - | 8 - 9 | 1198 | 61 | |
BC065071.1 | - | 10 - 11 | 2109 | 61 |