Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Plcxd1
- Entrez Gene ID
-
403178
- Gene Name
- phosphatidylinositol-specific phospholipase C, X domain containing 1
- Gene Aliases
-
A330045H12Rik, Gm576
- Chromosome Location
-
Chr.5: 110099225 - 110105953 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207279.2 |
NP_997162.2 |
1 - 2 |
52 |
85 |
View Full Product Details