Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldnd2
Entrez Gene ID
74276
Gene Name
claudin domain containing 2
Gene Aliases
1700071E18Rik
Chromosome Location
Chr.7: 43440816 - 43443320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028849.1 NP_083125.1 3 - 4 392 72
GenBank mRNA AK006929.1 - 3 - 4 427 72
BC128277.1 - 3 - 4 392 72

View Full Product Details