The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028236.1 | NP_082512.1 | 3 - 4 | 704 | 79 |
GenBank mRNA | AK008389.1 | - | 3 - 4 | 736 | 79 |
BC042780.1 | - | 3 - 4 | 704 | 79 |