Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sez6l
Entrez Gene ID
56747
Gene Name
seizure related 6 homolog like
Gene Aliases
AI843918, AIG1, AW539724, Acig1, BSRP-B, mKIAA0927
Chromosome Location
Chr.5: 112419151 - 112578042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253916.1 NP_001240845.1 1 - 2 434 85
NM_001253917.1 NP_001240846.1 1 - 2 434 85
NM_019982.3 NP_064366.2 1 - 2 434 85
GenBank mRNA AB206790.1 - 1 - 2 413 85
AK083229.1 - 1 - 2 398 85
BC065117.1 - 1 - 2 394 85
BC065129.1 - 1 - 2 211 85
CJ115240.1 - 1 - 2 434 85
DQ167195.1 - 1 - 2 421 85

View Full Product Details