Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptpn5
Entrez Gene ID
19259
Gene Name
protein tyrosine phosphatase, non-receptor type 5
Gene Aliases
Step
Chromosome Location
Chr.7: 47077795 - 47133695 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163565.1 NP_001157037.1 12 - 13 1795 78
NM_013643.2 NP_038671.2 12 - 13 1903 78
GenBank mRNA AK038416.1 - 12 - 13 1572 78
BC051975.1 - 12 - 13 1544 78
BC057339.1 - 11 - 12 2065 78
BC079592.1 - 12 - 13 1875 78
U28217.1 - 12 - 13 1914 78

View Full Product Details