The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163565.1 | NP_001157037.1 | 7 - 8 | 1213 | 83 |
NM_013643.2 | NP_038671.2 | 7 - 8 | 1321 | 83 | |
GenBank mRNA | AK038416.1 | - | 7 - 8 | 990 | 83 |
BC051975.1 | - | 7 - 8 | 962 | 83 | |
BC057339.1 | - | 6 - 7 | 1483 | 83 | |
BC079592.1 | - | 7 - 8 | 1293 | 83 | |
S80329.1 | - | 4 - 5 | 1157 | 83 | |
U28216.1 | - | 7 - 8 | 1074 | 83 | |
U28217.1 | - | 7 - 8 | 1332 | 83 |