Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc2
Entrez Gene ID
70546
Gene Name
zinc finger, DHHC domain containing 2
Gene Aliases
5730415P04Rik, 6430583A19Rik
Chromosome Location
Chr.8: 40423000 - 40489592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178395.3 NP_848482.1 8 - 9 961 92
GenBank mRNA AK018284.1 - - 933 92
AK030662.1 - 8 - 9 962 92
AK046533.1 - 8 - 9 961 92
AK136296.1 - 8 - 9 960 92
BC117761.1 - 9 - 10 723 92
BC127156.1 - 9 - 10 795 92

View Full Product Details