Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efr3a
Entrez Gene ID
76740
Gene Name
EFR3 homolog A (S. cerevisiae)
Gene Aliases
A130089M23Rik, BB071175, C76891, C920006C10Rik, D030063F01Rik, mKIAA0143
Chromosome Location
Chr.15: 65787020 - 65873812 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133766.3 NP_598527.2 11 - 12 1464 104
GenBank mRNA AB158474.1 - 11 - 12 1468 104
AK030074.1 - 11 - 12 1465 104
AK032461.1 - 11 - 12 1437 104
AK044000.1 - 11 - 12 1458 104
AK044475.1 - 11 - 12 1460 104
AK129065.1 - 11 - 12 1442 104
AK161250.1 - 11 - 12 1449 104
BC007482.1 - 11 - 12 1468 104
BC032092.1 - 11 - 12 1469 104

View Full Product Details