Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnah10
Entrez Gene ID
56087
Gene Name
dynein, axonemal, heavy chain 10
Gene Aliases
Dnahc10
Chromosome Location
Chr.5: 124725018 - 124834827 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019536.1 NP_062409.1 78 - 79 13706 98
GenBank mRNA AK147443.1 - 30 - 31 7331 98

View Full Product Details