Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt3
Entrez Gene ID
20981
Gene Name
synaptotagmin III
Gene Aliases
AI385753, sytIII
Chromosome Location
Chr.7: 44384099 - 44400030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016663.3 NP_057872.3 2 - 3 196 73
GenBank mRNA BY293405.1 - 2 - 3 169 73
D45858.1 - 1 - 2 130 73

View Full Product Details