Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acpt
Entrez Gene ID
100503991
Gene Name
acid phosphatase, testicular
Gene Aliases
EG546967, Gm1432
Chromosome Location
Chr.7: 44251761 - 44261396 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195034.1 NP_001181963.1 2 - 3 217 60

View Full Product Details