Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shank1
Entrez Gene ID
243961
Gene Name
SH3/ankyrin domain gene 1
Gene Aliases
-
Chromosome Location
Chr.7: 44307848 - 44358353 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034115.1 NP_001029287.1 12 - 13 1822 127

View Full Product Details