The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178701.3 | NP_848816.3 | 1 - 2 | 280 | 73 |
GenBank mRNA | AK050400.1 | - | 1 - 2 | 250 | 73 |
AK158262.1 | - | 1 - 2 | 281 | 73 | |
BC037717.1 | - | 1 - 2 | 261 | 73 |