Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uhrf2
Entrez Gene ID
109113
Gene Name
ubiquitin-like, containing PHD and RING finger domains 2
Gene Aliases
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf
Chromosome Location
Chr.19: 30030433 - 30093729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144873.2 NP_659122.2 12 - 13 2172 77
GenBank mRNA AB116653.1 - 12 - 13 1907 77
AF274047.1 - 12 - 13 2082 77
AK075912.1 - 1 - 2 117 77
AK170648.1 - 3 - 4 328 77
BC060241.1 - 12 - 13 2172 77

View Full Product Details