The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134209.1 | NP_598970.1 | 1 - 1 | 726 | 120 |
GenBank mRNA | BC125369.1 | - | 1 - 1 | 773 | 120 |
BC138162.1 | - | 1 - 1 | 773 | 120 |