Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stag3
Entrez Gene ID
50878
Gene Name
stromal antigen 3
Gene Aliases
SA-2
Chromosome Location
Chr.5: 138272589 - 138312397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016964.2 NP_058660.2 23 - 24 2589 67
GenBank mRNA AJ005678.1 - 23 - 24 2547 67
AK145898.1 - 20 - 21 2823 67
BC060184.1 - 22 - 23 3030 67
BC139010.1 - 23 - 24 2519 67
BC139011.1 - 23 - 24 2519 67

View Full Product Details