Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx25
Entrez Gene ID
102141
Gene Name
sorting nexin 25
Gene Aliases
AI661919, Gm1699, Gm173, Sbbi31
Chromosome Location
Chr.8: 46033261 - 46151858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207213.2 NP_997096.2 15 - 16 2228 69
GenBank mRNA AY601646.1 - 14 - 15 2447 69
BC054725.1 - 10 - 11 1484 69
BC057063.1 - 14 - 15 1976 69

View Full Product Details