Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Neil3
Entrez Gene ID
234258
Gene Name
nei like 3 (E. coli)
Gene Aliases
AI449477, BC034753, C85903
Chromosome Location
Chr.8: 53586867 - 53639065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146208.2 NP_666320.1 5 - 6 862 89
GenBank mRNA AB072931.1 - 5 - 6 729 89
BC024921.1 - 5 - 6 721 89
BC034753.1 - 5 - 6 805 89
DV646606.1 - 3 - 4 309 89

View Full Product Details