Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam71e1
Entrez Gene ID
75538
Gene Name
family with sequence similarity 71, member E1
Gene Aliases
0610007G24Rik, 1700021N13Rik, 1700021P22Rik, GARI-L5
Chromosome Location
Chr.7: 44496584 - 44501681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028169.1 NP_082445.1 2 - 3 266 71
GenBank mRNA AB473729.1 - 2 - 3 166 71
AK006229.1 - 1 - 2 384 71
BC130017.1 - 2 - 3 209 71

View Full Product Details