The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028169.1 | NP_082445.1 | 4 - 5 | 684 | 132 |
GenBank mRNA | AB473729.1 | - | 4 - 5 | 584 | 132 |
BC049752.1 | - | 4 - 5 | 632 | 132 | |
BC100470.1 | - | 4 - 5 | 635 | 132 | |
BC130017.1 | - | 4 - 5 | 627 | 132 |