The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285517.1 | NP_001272446.1 | 4 - 5 | 460 | 76 |
NM_009473.3 | NP_033499.1 | 4 - 5 | 467 | 76 | |
GenBank mRNA | BC066025.1 | - | 4 - 5 | 418 | 76 |
BQ573162.1 | - | 4 - 5 | 437 | 76 | |
U09419.1 | - | 3 - 4 | 285 | 76 |