Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfc5
Entrez Gene ID
72151
Gene Name
replication factor C (activator 1) 5
Gene Aliases
2610020K06Rik, 2610209F07Rik, 36.5kDa, 36kDa, Recc5
Chromosome Location
Chr.5: 117379145 - 117389023 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028128.1 NP_082404.1 3 - 4 315 113
GenBank mRNA AK011489.1 - 3 - 4 326 113
AK150036.1 - - 328 113
AK170211.1 - 3 - 4 333 113
AK171372.1 - - 315 113
BC023674.1 - 2 - 3 253 113
BC089001.1 - 3 - 4 315 113

View Full Product Details