Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spcs3
Entrez Gene ID
76687
Gene Name
signal peptidase complex subunit 3 homolog (S. cerevisiae)
Gene Aliases
1810011E08Rik
Chromosome Location
Chr.8: 54520433 - 54530739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029701.1 NP_083977.1 1 - 2 233 85
GenBank mRNA AK007432.1 - 1 - 2 212 85
AK028314.1 - 1 - 2 195 85
AK048051.1 - 1 - 2 219 85
BC019390.1 - 1 - 2 172 85
BC054817.1 - 1 - 2 198 85
BY012914.1 - 1 - 2 233 85

View Full Product Details