The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253843.1 | NP_001240772.1 | 7 - 8 | 842 | 92 |
NM_001253844.1 | NP_001240773.1 | 5 - 6 | 435 | 92 | |
NM_134129.4 | NP_598890.1 | 7 - 8 | 785 | 92 | |
GenBank mRNA | AB222602.1 | - | 7 - 8 | 92 | |
AF251503.1 | - | 7 - 8 | 750 | 92 | |
AK047693.1 | - | 7 - 8 | 660 | 92 | |
AK076998.1 | - | 5 - 6 | 436 | 92 | |
AK088769.1 | - | 7 - 8 | 770 | 92 | |
AK164681.1 | - | 7 - 8 | 773 | 92 | |
BC004070.1 | - | 7 - 8 | 92 |