Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink4
Entrez Gene ID
20731
Gene Name
serine peptidase inhibitor, Kazal type 4
Gene Aliases
MPGC60
Chromosome Location
Chr.4: 40920056 - 40931395 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011463.2 NP_035593.2 1 - 2 96 60
GenBank mRNA BC117048.1 - 1 - 2 99 60
BC117050.1 - 1 - 2 98 60
CF381698.1 - 1 - 2 102 60

View Full Product Details